siRNA Details
| Gene Symbol: Tnrc6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198022.2 | NP_932139.2 | 5 | 1460 | ||
| GenBank mRNA | AK147471.1 | - | 6 | 1458 | ||
| AK147551.1 | - | 5 | 1176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnrc6c| Gene Symbol: Tnrc6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198022.2 | NP_932139.2 | 5 | 1460 | ||
| GenBank mRNA | AK147471.1 | - | 6 | 1458 | ||
| AK147551.1 | - | 5 | 1176 | |||