siRNA Details
| Gene Symbol: Nploc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195023.1 | NP_001181952.1 | 14, 15 | 1647 | ||
| NM_199469.2 | NP_955763.1 | 13, 14 | 1551 | |||
| GenBank mRNA | AK129375.1 | - | 14, 15 | 1622 | ||
| AK141998.1 | - | 14, 15 | 1646 | |||
| AK149823.1 | - | 14, 15 | 1619 | |||
| AK149912.1 | - | 14, 15 | 1646 | |||
| AK215926.1 | - | 1, 2 | 35 | |||
| BC065156.1 | - | 13, 14 | 1519 | 5706754 | ||

Nploc4
Genomic Map