siRNA Details
| Gene Symbol: Rab40b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139147.3 | NP_631886.2 | 5 | 589 | ||
| GenBank mRNA | AA718654.1 | - | 2 | 175 | ||
| AB232638.1 | - | 5 | 477 | |||
| AF425643.1 | - | 5 | 477 | |||
| BC096601.1 | - | 5 | 583 | 4974190 | ||

Rab40b
Genomic Map