siRNA Details
| Gene Symbol: Trib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144551.5 | NP_653134.2 | 3 | 1942 | ||
| GenBank mRNA | AF358867.1 | - | 3 | 807 | ||
| AK044747.1 | - | 3 | 1900 | |||
| AK080064.1 | - | 3 | 1145 | |||
| AK082329.1 | - | 3 | 1154 | |||
| AK144192.1 | - | 3 | 2007 | |||
| BC027159.1 | - | 2 | 399 | |||
| BC034338.1 | - | 3 | 976 | 5043179 | ||
| BC037387.1 | - | 3 | 1886 | 5363174 | ||

Trib2
Genomic Map