siRNA Details
| Gene Symbol: Nol10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008421.1 | NP_001008421.1 | 12, 13 | 1044 | ||
| GenBank mRNA | AK148210.1 | - | 12, 13 | 1060 | ||
| AK160864.1 | - | 12, 13 | 1080 | |||
| AK164400.1 | - | 12, 13 | 1070 | |||
| AK166160.1 | - | 12, 13 | 1073 | |||
| BC086676.1 | - | 12, 13 | 1044 | 30659737 | ||

Nol10
Genomic Map