siRNA Details
| Gene Symbol: Adad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009350.3 | NP_033376.2 | 11 | 1736 | ||
| GenBank mRNA | AK133008.1 | - | 8 | 1152 | ||
| X84693.1 | - | 11 | 1724 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adad1| Gene Symbol: Adad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009350.3 | NP_033376.2 | 11 | 1736 | ||
| GenBank mRNA | AK133008.1 | - | 8 | 1152 | ||
| X84693.1 | - | 11 | 1724 | |||