siRNA Details
Gene Symbol: Adad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009350.3 | NP_033376.2 | 11, 12 | 1817 | ||
GenBank mRNA | AK133008.1 | - | 8, 9 | 1233 | ||
X84693.1 | - | 11, 12 | 1805 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009350.3 | NP_033376.2 | 11, 12 | 1817 | ||
GenBank mRNA | AK133008.1 | - | 8, 9 | 1233 | ||
X84693.1 | - | 11, 12 | 1805 |