siRNA Details
| Gene Symbol: Ankmy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146033.3 | NP_666145.3 | 5 | 683 | ||
| GenBank mRNA | AK037644.1 | - | 3 | 3442 | ||
| AK077556.1 | - | 5 | 654 | |||
| AK164434.1 | - | 5 | 585 | |||
| BC012275.1 | - | 5 | 649 | 4010781 | ||
| BC024959.1 | - | 5 | 640 | 4946879 | ||

Ankmy2
Genomic Map