siRNA Details
| Gene Symbol: Ankmy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146033.3 | NP_666145.3 | 5, 6 | 762 | ||
| GenBank mRNA | AK037644.1 | - | 3, 4 | 3521 | ||
| AK077556.1 | - | 5, 6 | 733 | |||
| AK164434.1 | - | 5, 6 | 664 | |||
| BC012275.1 | - | 5, 6 | 728 | 4010781 | ||
| BC024959.1 | - | 5, 6 | 719 | 4946879 | ||

Ankmy2
Genomic Map