siRNA Details
| Gene Symbol: Tert | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009354.1 | NP_033380.1 | 2 | 1088 | ||
| GenBank mRNA | AF051911.1 | - | 2 | 1088 | ||
| AF073311.1 | - | 2 | 1059 | |||
| AK133783.1 | - | 2 | 1179 | |||
| BC082327.1 | - | 3 | 590 | |||
| BC127068.1 | - | 3 | 569 | 40110173 | ||

Tert
Genomic Map