siRNA Details
| Gene Symbol: G2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015099.2 | NP_001015099.2 | 4, 5 | 455 | ||
| NM_001167963.1 | NP_001161435.1 | 4, 5 | 455 | |||
| NM_001167964.1 | NP_001161436.1 | 4, 5 | 455 | |||
| GenBank mRNA | AK031333.1 | - | 4, 5 | 315 | ||
| AK033513.1 | - | 4, 5 | 397 | |||
| AK084256.1 | - | 4, 5 | 326 | |||
| AK129332.1 | - | 4, 5 | 358 | |||
| BC086455.1 | - | 4, 5 | 328 | 30458160 | ||

G2e3
Genomic Map