siRNA Details
| Gene Symbol: Slc25a21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167976.1 | NP_001161448.1 | 6, 7 | 424 | ||
| NM_172577.3 | NP_766165.2 | 5, 6 | 867 | |||
| GenBank mRNA | AK036988.1 | - | 5, 6 | 607 | ||
| AK041694.1 | - | 5, 6 | 868 | |||
| AK045180.1 | - | 5, 6 | 693 | |||
| AK162379.1 | - | 5, 6 | 694 | |||
| AK167535.1 | - | 5, 6 | 575 | |||
| BC057980.1 | - | 5, 6 | 691 | 5042653 | ||
| EU180236.1 | - | 6, 7 | 424 | |||

Slc25a21
Genomic Map