siRNA Details
| Gene Symbol: Tex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198292.3 | NP_938034.2 | 2 | 1499 | ||
| GenBank mRNA | AK029537.1 | - | 2 | 1506 | ||
| AK129434.1 | - | 2 | 1499 | |||
| AK168397.1 | - | 2 | 1509 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex2| Gene Symbol: Tex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198292.3 | NP_938034.2 | 2 | 1499 | ||
| GenBank mRNA | AK029537.1 | - | 2 | 1506 | ||
| AK129434.1 | - | 2 | 1499 | |||
| AK168397.1 | - | 2 | 1509 | |||