siRNA Details
| Gene Symbol: L2hgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145443.2 | NP_663418.1 | 8 | 1037 | ||
| GenBank mRNA | AK145228.1 | - | 8 | 1036 | ||
| AK152450.1 | - | 8 | 1014 | |||
| AK168429.1 | - | 8 | 985 | |||
| BC016226.1 | - | 8 | 981 | 4487011 | ||

L2hgdh
Genomic Map