siRNA Details
| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009357.2 | NP_033383.1 | 3, 4 | 401 | ||
| GenBank mRNA | AK149789.1 | - | 3, 4 | 366 | ||
| AK157834.1 | - | 3, 4 | 380 | |||
| AK190569.1 | - | 1, 2 | 35 | |||
| BC003802.1 | - | 3, 4 | 345 | 3486968 | ||
| BY250528.1 | - | 3, 4 | 402 | |||
| X81058.1 | - | 3, 4 | 347 | |||

Tex261
Genomic Map