siRNA Details
| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009357.2 | NP_033383.1 | 5, 6 | 571 | ||
| GenBank mRNA | AK149789.1 | - | 5, 6 | 536 | ||
| BC003802.1 | - | 5, 6 | 515 | 3486968 | ||
| X81058.1 | - | 5, 6 | 517 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex261| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009357.2 | NP_033383.1 | 5, 6 | 571 | ||
| GenBank mRNA | AK149789.1 | - | 5, 6 | 536 | ||
| BC003802.1 | - | 5, 6 | 515 | 3486968 | ||
| X81058.1 | - | 5, 6 | 517 | |||