siRNA Details
| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009357.2 | NP_033383.1 | 6 | 617 | ||
| GenBank mRNA | AK149789.1 | - | 6 | 582 | ||
| AK157834.1 | - | 4 | 1431 | |||
| AK188173.1 | - | 1 | 49 | |||
| AK218434.1 | - | 1 | 49 | |||
| BC003802.1 | - | 6 | 561 | 3486968 | ||
| X81058.1 | - | 6 | 563 | |||

Tex261
Genomic Map