siRNA Details
| Gene Symbol: Ppp2r5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009358.3 | NP_033384.2 | 7, 8 | 972 | ||
| GenBank mRNA | AB053464.1 | - | 7, 8 | 917 | ||
| AF316616.2 | - | 7, 8 | 829 | |||
| AK154050.1 | - | 7, 8 | 948 | |||
| AK160587.1 | - | 7, 8 | 913 | |||
| AY338234.1 | - | 7, 8 | 902 | |||
| BC010716.1 | - | 7, 8 | 885 | 2655861 | ||

Ppp2r5d
Genomic Map