siRNA Details
| Gene Symbol: Eif2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145445.3 | NP_663420.1 | 4 | 604 | ||
| GenBank mRNA | AK049731.1 | - | 4 | 605 | ||
| AK076195.1 | - | 4 | 600 | |||
| BC003326.1 | - | 4 | 636 | 3156632 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2b2| Gene Symbol: Eif2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145445.3 | NP_663420.1 | 4 | 604 | ||
| GenBank mRNA | AK049731.1 | - | 4 | 605 | ||
| AK076195.1 | - | 4 | 600 | |||
| BC003326.1 | - | 4 | 636 | 3156632 | ||