siRNA Details
Gene Symbol: Eif2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145445.3 | NP_663420.1 | 4, 5 | 653 | ||
GenBank mRNA | AK049731.1 | - | 4, 5 | 654 | ||
AK076195.1 | - | 4, 5 | 649 | |||
AK194280.1 | - | 1, 2 | 45 | |||
BC003326.1 | - | 4, 5 | 685 | 3156632 |
