siRNA Details
| Gene Symbol: Cirh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011574.2 | NP_035704.2 | 15 | 1813 | ||
| GenBank mRNA | AK144455.1 | - | 15 | 1798 | ||
| AK146263.1 | - | 15 | 1789 | |||
| AK150862.1 | - | 15 | 1788 | |||
| AK168470.1 | - | 14 | 2705 | |||
| AK173318.1 | - | 14 | 1754 | |||
| BC018262.1 | - | 3 | 266 | |||
| BC027399.1 | - | 11 | 1312 | 4948207 | ||
| BC068113.1 | - | 15 | 1802 | 5707865 | ||

Cirh1a
Genomic Map