siRNA Details
| Gene Symbol: Itpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172584.3 | NP_766172.1 | 6, 7 | 550 | ||
| GenBank mRNA | AK038913.1 | - | 6, 7 | 551 | ||
| BC056464.1 | - | 6, 7 | 531 | 5707377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpk1| Gene Symbol: Itpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172584.3 | NP_766172.1 | 6, 7 | 550 | ||
| GenBank mRNA | AK038913.1 | - | 6, 7 | 551 | ||
| BC056464.1 | - | 6, 7 | 531 | 5707377 | ||