siRNA Details
| Gene Symbol: Tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019678.3 | NP_062652.1 | 4 | 670 | ||
| GenBank mRNA | AK008549.1 | - | 4 | 446 | ||
| AK158539.1 | - | 4 | 536 | |||
| AK167475.1 | - | 4 | 534 | |||
| BC024638.1 | - | 4 | 448 | 4015300 | ||
| U94662.1 | - | 4 | 572 | |||

Tfg
Genomic Map