siRNA Details
| Gene Symbol: Rapgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175930.5 | NP_787126.3 | 9 | 1204 | ||
| GenBank mRNA | AK029995.1 | - | 9 | 1205 | ||
| AK030016.1 | - | 9 | 1158 | |||
| AK162220.1 | - | 7 | 837 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rapgef5| Gene Symbol: Rapgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175930.5 | NP_787126.3 | 9 | 1204 | ||
| GenBank mRNA | AK029995.1 | - | 9 | 1205 | ||
| AK030016.1 | - | 9 | 1158 | |||
| AK162220.1 | - | 7 | 837 | |||