siRNA Details
| Gene Symbol: Rapgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175930.5 | NP_787126.3 | 28 | 3001 | ||
| GenBank mRNA | AK029995.1 | - | 28 | 3002 | ||
| AK030016.1 | - | 28 | 2955 | |||
| AK083591.1 | - | 18 | 1941 | |||
| AK122234.1 | - | 16 | 1763 | |||
| BC046627.1 | - | 16 | 1722 | 6494175 | ||

Rapgef5
Genomic Map