siRNA Details
Gene Symbol: Cdca7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146040.1 | NP_666152.1 | 2 | 111 | ||
GenBank mRNA | AK149673.1 | - | 2 | 89 | ||
BC006933.1 | - | 2 | 111 | 3602150 | ||
BC071198.1 | - | 2 | 78 |
