siRNA Details
| Gene Symbol: Cdca7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146040.1 | NP_666152.1 | 4 | 614 | ||
| GenBank mRNA | AK149673.1 | - | 4 | 592 | ||
| BC006933.1 | - | 4 | 614 | 3602150 | ||
| BC071198.1 | - | 4 | 582 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdca7l| Gene Symbol: Cdca7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146040.1 | NP_666152.1 | 4 | 614 | ||
| GenBank mRNA | AK149673.1 | - | 4 | 592 | ||
| BC006933.1 | - | 4 | 614 | 3602150 | ||
| BC071198.1 | - | 4 | 582 | |||