siRNA Details
| Gene Symbol: Larp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172585.2 | NP_766173.1 | 11 | 1329 | ||
| GenBank mRNA | AK042491.1 | - | 11 | 1327 | ||
| AK080953.1 | - | 11 | 1353 | |||
| AK160759.1 | - | 11 | 1330 | |||
| AK172916.1 | - | 11 | 1311 | |||
| BC059218.1 | - | 11 | 1308 | |||
| BC078640.1 | - | 11 | 1252 | 6506267 | ||
| BC086626.1 | - | 11 | 1308 | |||

Larp4b
Genomic Map