siRNA Details
| Gene Symbol: Larp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172585.2 | NP_766173.1 | 17, 18 | 2238 | ||
| GenBank mRNA | AK080953.1 | - | 17, 18 | 2262 | ||
| AK172916.1 | - | 17, 18 | 2220 | |||
| BC059218.1 | - | 17, 18 | 2217 | |||
| BC078640.1 | - | 16, 17 | 1909 | 6506267 | ||
| BC086626.1 | - | 17, 18 | 2217 | |||

Larp4b
Genomic Map