siRNA Details
| Gene Symbol: Larp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172585.2 | NP_766173.1 | 15 | 1994 | ||
| GenBank mRNA | AK042491.1 | - | 15 | 1992 | ||
| AK080953.1 | - | 15 | 2018 | |||
| AK172916.1 | - | 15 | 1976 | |||
| BC059218.1 | - | 15 | 1973 | |||
| BC078640.1 | - | 14 | 1665 | 6506267 | ||
| BC086626.1 | - | 15 | 1973 | |||

Larp4b
Genomic Map