siRNA Details
| Gene Symbol: Heatr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144835.4 | NP_659084.3 | 33, 34 | 4764 | ||
| GenBank mRNA | AK132192.1 | - | 438 | |||
| BQ917385.1 | - | 4, 5 | 516 | |||
| CF724982.1 | - | 2, 3 | 161 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heatr1| Gene Symbol: Heatr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144835.4 | NP_659084.3 | 33, 34 | 4764 | ||
| GenBank mRNA | AK132192.1 | - | 438 | |||
| BQ917385.1 | - | 4, 5 | 516 | |||
| CF724982.1 | - | 2, 3 | 161 | |||