siRNA Details
| Gene Symbol: Amph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289546.1 | NP_001276475.1 | 18 | 1628 | ||
| NM_175007.2 | NP_778172.1 | 18 | 1616 | |||
| GenBank mRNA | AK132168.1 | - | 18 | 1629 | ||
| BC024817.1 | - | 8 | 597 | |||
| BC043718.1 | - | 2 | 285 | |||
| BC051425.1 | - | 4 | 2938 | |||
| BC054718.1 | - | 18 | 1596 | 5686478 | ||

Amph
Genomic Map