siRNA Details
Gene Symbol: Amph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289546.1 | NP_001276475.1 | 3, 4 | 276 | ||
NM_175007.2 | NP_778172.1 | 3, 4 | 276 | |||
GenBank mRNA | AK047144.1 | - | 3, 4 | 230 | ||
AK132168.1 | - | 3, 4 | 277 | |||
BC054718.1 | - | 3, 4 | 256 | 5686478 |
