siRNA Details
| Gene Symbol: Tgfb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009367.3 | NP_033393.2 | 2 | 1670 | ||
| GenBank mRNA | AK141552.1 | - | 3 | 1052 | ||
| AK159690.1 | - | 2 | 234 | |||
| BC011170.1 | - | 2 | 578 | 4035366 | ||
| X57413.1 | - | 2 | 1669 | |||

Tgfb2
Genomic Map