siRNA Details
| Gene Symbol: Slc17a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144836.2 | NP_659085.1 | 8 | 1313 | ||
| GenBank mRNA | AK040717.1 | - | 8 | 1314 | ||
| AK050184.1 | - | 8 | 1296 | |||
| AK050277.1 | - | 8 | 1190 | |||
| BC018306.1 | - | 7 | 951 | 4193755 | ||

Slc17a2
Genomic Map