siRNA Details
| Gene Symbol: Slc17a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144836.2 | NP_659085.1 | 5 | 898 | ||
| GenBank mRNA | AK040717.1 | - | 5 | 899 | ||
| AK050184.1 | - | 5 | 881 | |||
| AK050277.1 | - | 6 | 862 | |||
| BC018306.1 | - | 4 | 536 | 4193755 | ||

Slc17a2
Genomic Map