siRNA Details
| Gene Symbol: Thbd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009378.3 | NP_033404.1 | 1 | 1579 | ||
| GenBank mRNA | AK044928.1 | - | 1 | 1511 | ||
| AK089479.1 | - | 1 | 1580 | |||
| X14432.1 | - | 1 | 1495 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thbd| Gene Symbol: Thbd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009378.3 | NP_033404.1 | 1 | 1579 | ||
| GenBank mRNA | AK044928.1 | - | 1 | 1511 | ||
| AK089479.1 | - | 1 | 1580 | |||
| X14432.1 | - | 1 | 1495 | |||