siRNA Details
Gene Symbol: Thbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011582.3 | NP_035712.1 | 16 | 2124 | ||
GenBank mRNA | AF102887.1 | - | 16 | 2064 | ||
BC139414.1 | - | 16 | 2004 | 8862436 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Thbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011582.3 | NP_035712.1 | 16 | 2124 | ||
GenBank mRNA | AF102887.1 | - | 16 | 2064 | ||
BC139414.1 | - | 16 | 2004 | 8862436 |