siRNA Details
| Gene Symbol: Ttc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081352.1 | NP_001074821.1 | 33 | 3706 | ||
| GenBank mRNA | AK129128.1 | - | 12 | 1062 | ||
| BC035338.1 | - | 14 | 1994 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc37| Gene Symbol: Ttc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081352.1 | NP_001074821.1 | 33 | 3706 | ||
| GenBank mRNA | AK129128.1 | - | 12 | 1062 | ||
| BC035338.1 | - | 14 | 1994 | |||