siRNA Details
| Gene Symbol: Rasa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145452.3 | NP_663427.2 | 8 | 1783 | ||
| GenBank mRNA | AK042801.1 | - | 1 | 1142 | ||
| AK133354.1 | - | 4 | 343 | |||
| BC013637.1 | - | 6 | 529 | 3498774 | ||
| BC065082.1 | - | 8 | 1326 | |||

Rasa1
Genomic Map