siRNA Details
| Gene Symbol: Tia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164078.1 | NP_001157550.1 | 8, 9 | 924 | ||
| NM_001164079.1 | NP_001157551.1 | 8, 9 | 918 | |||
| NM_011585.4 | NP_035715.1 | 9, 10 | 951 | |||
| GenBank mRNA | AK028340.1 | - | 7, 8 | 591 | ||
| AK088486.1 | - | 9, 10 | 887 | |||
| AK144222.1 | - | 9, 10 | 904 | |||
| BC023813.1 | - | 8, 9 | 833 | 5353938 | ||
| BC027826.1 | - | 7, 8 | 2888 | |||
| BC046812.1 | - | 8, 9 | 807 | 5702183 | ||
| U00689.1 | - | 9, 10 | 892 | |||
| U55862.1 | - | 9, 10 | 759 | |||

Tia1
Genomic Map