siRNA Details
| Gene Symbol: Srek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172592.2 | NP_766180.1 | 2 | 748 | ||
| GenBank mRNA | AK033354.1 | - | 2 | 748 | ||
| AK037115.1 | - | 2 | 724 | |||
| AK039311.1 | - | 4 | 483 | |||
| AK044086.1 | - | 1 | 52 | |||
| AK052265.1 | - | 4 | 617 | |||
| AK054414.1 | - | 2 | 1199 | |||
| BC063761.1 | - | 5 | 493 | |||
| BC070460.1 | - | 2 | 121 | 30548562 | ||

Srek1
Genomic Map