siRNA Details
| Gene Symbol: Paip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079849.2 | NP_001073318.1 | 4 | 494 | ||
| NM_145457.4 | NP_663432.1 | 4 | 712 | |||
| GenBank mRNA | AF128835.1 | - | 3 | 394 | ||
| AK040012.1 | - | 4 | 495 | |||
| BB202059.2 | - | 1 | 83 | |||
| BC019726.1 | - | 4 | 683 | 4010170 | ||
| BC051042.1 | - | 4 | 669 | 4008643 | ||
| CB840007.1 | - | 3 | 467 | |||
| CD363907.1 | - | 4 | 470 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108937, XM_008760778, XM_008760780, XM_008760779, BC166996.

Paip1
Genomic Map