siRNA Details
| Gene Symbol: Rarb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289760.1 | NP_001276689.1 | 9 | 1628 | ||
| NM_001289761.1 | NP_001276690.1 | 8 | 1547 | |||
| NM_001289762.1 | NP_001276691.1 | 7 | 1157 | |||
| NM_011243.2 | NP_035373.1 | 7 | 1511 | |||
| GenBank mRNA | AK043726.1 | - | 7 | 1727 | ||
| AK078471.1 | - | 7 | 1223 | |||
| BC076597.1 | - | 7 | 1507 | 30608242 | ||
| S56660.1 | - | 7 | 1407 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK140978.1 | - | 1303 | Chr. - - [-] on Build | ||

Rarb
Genomic Map