siRNA Details
| Gene Symbol: Tll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009390.2 | NP_033416.2 | 20, 21 | 3525 | ||
| GenBank mRNA | AK139103.1 | - | 8, 9 | 1314 | ||
| U34042.1 | - | 20, 21 | 3525 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tll1| Gene Symbol: Tll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009390.2 | NP_033416.2 | 20, 21 | 3525 | ||
| GenBank mRNA | AK139103.1 | - | 8, 9 | 1314 | ||
| U34042.1 | - | 20, 21 | 3525 | |||