siRNA Details
Gene Symbol: Tln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011602.5 | NP_035732.2 | 45, 46 | 6544 | ||
GenBank mRNA | AK122420.2 | - | 45, 46 | 6170 | ||
AK147228.1 | - | 45, 46 | 6115 | |||
AK170630.1 | - | 11, 12 | 1587 | |||
AK171322.1 | - | 11, 12 | 1587 | |||
BC018557.1 | - | 9, 10 | 1252 | |||
BC121787.1 | - | 15, 16 | 2056 | |||
BC150810.1 | - | 45, 46 | 6215 | 9087721 | ||
X56123.1 | - | 45, 46 | 6216 |
