siRNA Details
Gene Symbol: Tln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011602.5 | NP_035732.2 | 35 | 5090 | ||
GenBank mRNA | AK122420.2 | - | 35 | 4716 | ||
AK147228.1 | - | 35 | 4661 | |||
AK170630.1 | - | 1 | 133 | |||
AK171322.1 | - | 1 | 133 | |||
BC121787.1 | - | 5 | 602 | |||
BC150810.1 | - | 35 | 4761 | 9087721 | ||
X56123.1 | - | 35 | 4762 |
