siRNA Details
| Gene Symbol: Atg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172599.4 | NP_766187.1 | 8 | 1075 | ||
| GenBank mRNA | AK029330.1 | - | 8 | 1055 | ||
| AK029967.1 | - | 8 | 1056 | |||
| AK173037.1 | - | 2 | 129 | |||
| BC025038.1 | - | 1 | 66 | |||
| BC057360.1 | - | 7 | 795 | |||
| BC090995.1 | - | 8 | 1055 | 6754513 | ||

Atg14
Genomic Map