siRNA Details
| Gene Symbol: Atg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172599.4 | NP_766187.1 | 8, 9 | 1108 | ||
| GenBank mRNA | AK029330.1 | - | 8, 9 | 1088 | ||
| AK029967.1 | - | 8, 9 | 1089 | |||
| AK173037.1 | - | 2, 3 | 162 | |||
| BC025038.1 | - | 1, 2 | 99 | |||
| BC057360.1 | - | 7, 8 | 828 | |||
| BC090995.1 | - | 8, 9 | 1088 | 6754513 | ||

Atg14
Genomic Map