siRNA Details
| Gene Symbol: Tlr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021297.2 | NP_067272.1 | 3 | 1996 | ||
| GenBank mRNA | AF095353.1 | - | 3 | 2182 | ||
| AF110133.1 | - | 3 | 1996 | |||
| AF185285.1 | - | 3 | 1975 | |||
| AK030348.1 | - | 3 | 2196 | |||
| AK154297.1 | - | 3 | 2089 | |||
| AK170566.1 | - | 3 | 2048 | |||
| BC029856.1 | - | 3 | 2208 | 3493732 | ||
| JX878358.1 | - | 3 | 1990 | |||
| JX878359.1 | - | 3 | 1990 | |||

Tlr4
Genomic Map